3-Bromo-L-phenylalanine 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(S)-2-Amino-3-(3-bromophenyl)propanoic acid
, H-Phe(3-Br)-OH
APOSOR925479-5GEA
33.7
GBP
APOSOR925479-5G
3-Bromo-L-phenylalanine 97%
3-Bromo-L-phenylalanine
Formula:
C₉H₁₀BrNO₂ MW: 244.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659110 CAS Number: 82311-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...