Methyl salicylate 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Methyl 2-hydroxybenzoate
, Wintergreen Oil
APOSOR924591-100GEA
28.5
GBP
APOSOR924591-100G
Methyl salicylate 95%
Methyl salicylate
Formula:
2-(HO)C₆H₄CO₂CH₃ MW: 152.15 g/mol Boiling Pt: 222 °C Density: 1.174 g/cm³ (25 °C) Flash Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002214 CAS Number: 119-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...