γ-Methyl-γ-decanolactone 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
γ-Methyl-γ-decalactone
OR924420-100G
OR924420-25G
APOSOR924420-100GEA
263
GBP
APOSOR924420-100G
APOSOR924420-25G
γ-Methyl-γ-decanolactone 95%
gamma-Methyl-gamma-decanolactone
Formula:
C₁₁H₂₀O₂ MW: 184.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036642 CAS Number: 7011-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...