N-Methylaniline 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR922839-100G OR922839-1KG OR922839-500G
APOSOR922839-100GEA 78.8 GBP
APOSOR922839-100G APOSOR922839-1KG APOSOR922839-500G
N-Methylaniline 95%
N-Methylaniline
Formula: C₇H₉N
MW: 107.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008283
CAS Number: 100-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR