Thymol 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Isopropyl-m-cresol2-Isopropyl-5-methylphenol4-Isopropyl-3-hydroxytoluene

APOSOR921899-100GEA 89.8 GBP
APOSOR921899-100G
Thymol 95%
Thymol
Formula: 2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
MW: 150.22 g/mol
Boiling Pt: 232 °C
Melting Pt: 48…51 °C
Density: 0.965 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002309
CAS Number: 89-83-8
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR