Chlorobenzene
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Benzene Chloride
, Phenylchloride
, Monochlorobenzene
APOSOR8791-1LEA
52.6
GBP
APOSOR8791-1L
Chlorobenzene
Chlorobenzene
Formula:
C₆H₅Cl MW: 112.56 g/mol Boiling Pt: 132 °C Melting Pt: –45 °C Density: 1.106 g/cm³ (25 °C) Flash Pt: 27 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000530 CAS Number: 108-90-7 EINECS: 203-628-5 UN: 1134 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...