4-Cyclohexylphenol 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Hydroxyphenylcyclohexane
APOSOR6485-100GEA
57.9
GBP
APOSOR6485-100G
APOSOR6485-5G
APOSOR6485-25G
APOSOR6485-10G
4-Cyclohexylphenol 97%
4-Cyclohexylphenol
Formula:
C₁₂H₁₆O MW: 176.26 g/mol Melting Pt: 127…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035699 CAS Number: 1131-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...