D,L-Anatabine 94%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: (R,S)-Anatabine1,2,3,6-Tetrahydro-2,3'-bipyridine

APOSOR6370T-100MGEA 66.2 GBP
APOSOR6370T-100MG APOSOR6370T-250MG
D,L-Anatabine 94%
D,L-Anatabine

A minor component in tobacco.

Formula: C₁₀H₁₂N₂
MW: 160.22 g/mol
Boiling Pt: 116…117 °C/1mmHg
Density: 1.086 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078278
CAS Number: 2743-90-0
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR