Indole-4-carbonitrile
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1H-Indole-4-carbonitrile
, 4-Cyano-1H-indole
, 4-Cyanoindole
APOSOR59809-5GEA
10.6
GBP
APOSOR59809-5G
Indole-4-carbonitrile
Indole-4-carbonitrile
Formula:
C₉H₆N₂ MW: 142.16 g/mol Melting Pt: 117…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00152045 CAS Number: 16136-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...