2-Methyloxan-4-one

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Methyldihydro-2H-pyran-4(3H)-one, 2-Methyltetrahydro-4H-pyran-4-one

OR54543-1G OR54543-250MG OR54543-5G
APOSOR54543-1GEA 64.1 GBP
APOSOR54543-1G APOSOR54543-250MG APOSOR54543-5G
2-Methyloxan-4-one
2-Methyloxan-4-one
Formula: C₆H₁₀O₂
MW: 114.142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15071428
CAS Number: 1193-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR