2,2,6-Trimethyl-4H-1,3-dioxin-4-one 94%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,2,6-Trimethyl-1,3-dioxine-4-one

OR54478-100G OR54478-500G
APOSOR54478-100GEA 94.9 GBP
APOSOR54478-100G APOSOR54478-500G
2,2,6-Trimethyl-4H-1,3-dioxin-4-one 94%
2,2,6-Trimethyl-4H-1,3-dioxin-4-one
Formula: C₇H₁₀O₃
MW: 142.15 g/mol
Boiling Pt: 65…67 °C
Melting Pt: 7 °C
Density: 1.094 g/cm³ (25 °C)
Flash Pt: 14 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040468
CAS Number: 5394-63-8
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR