1-Chloro-2-nitrobenzene 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-Chloro-2-nitrobenzene, 2-Chloronitrobenzene

OR5399-1KG OR5399-250G OR5399-5KG
APOSOR5399-1KGEA 94.9 GBP
APOSOR5399-1KG APOSOR5399-250G APOSOR5399-5KG
1-Chloro-2-nitrobenzene 99%
1-Chloro-2-nitrobenzene
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 245…246 °C
Melting Pt: 31…34 °C
Density: 1.348 g/cm³ (25 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00007061
CAS Number: 88-73-3
EINECS: 201-854-9
UN: 1578
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR