2-(Tributylstannyl)pyrimidine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR52038-10G OR52038-1G OR52038-5G
APOSOR52038-10GEA 263 GBP
APOSOR52038-10G APOSOR52038-1G APOSOR52038-5G
2-(Tributylstannyl)pyrimidine
2-(Tributylstannyl)pyrimidine
Formula: C₁₆H₃₀N₂Sn
Density: 1.164 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01319085
CAS Number: 153435-63-3
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR