3-Bromobiphenyl (PBB No. 2) 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: PBB No. 2

OR51897-100G OR51897-25G
APOSOR51897-100GEA 89.8 GBP
APOSOR51897-100G APOSOR51897-25G
3-Bromobiphenyl (PBB No. 2) 97%
3-Bromobiphenyl (PBB No. 2)
Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 103…104 °C/0.2mmHg
Density: 1.398 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000082
CAS Number: 2113-57-7
EINECS: 218-304-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR