(1H-Indol-3-yl)acetic acid 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: IAA, 3-(Carboxymethyl)-1H-indole, 1H-Indol-3-yl-acetic acid, Heteroauxin

OR5064-100G OR5064-25G OR5064-500G
APOSOR5064-100GEA 40 GBP
APOSOR5064-100G APOSOR5064-25G APOSOR5064-500G
(1H-Indol-3-yl)acetic acid 99%
(1H-Indol-3-yl)acetic acid
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 165…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005636
CAS Number: 87-51-4
EINECS: 201-748-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR