Isonicotinaldehyde 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Pyridine-4-carboxaldehyde, 4-Pyridinecarboxaldehyde, 4-Pyridinecarbaldehyde

OR5048-100G OR5048-500G
APOSOR5048-100GEA 73.6 GBP
APOSOR5048-100G APOSOR5048-500G
Isonicotinaldehyde 98%
Isonicotinaldehyde
Formula: C₆H₅NO
MW: 107.11 g/mol
Boiling Pt: 71…71 °C/10mmHg
Melting Pt: –4 °C
Density: 1.137 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006425
CAS Number: 872-85-5
EINECS: 212-832-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR