Biphenyl-2-ol 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Phenylphenol, 2-Hydroxydiphenyl, 2-Hydroxybiphenyl

OR4926-500G OR4926-25G OR4926-1KG
APOSOR4926-500GEA 58.9 GBP
APOSOR4926-500G APOSOR4926-25G APOSOR4926-1KG
Biphenyl-2-ol 99%
Biphenyl-2-ol
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 282…287 °C
Melting Pt: 54…59 °C
Density: 1.293 g/cm³ (25 °C)
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00002208
CAS Number: 90-43-7
EINECS: 201-993-5
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR