N-Fmoc-L-norleucine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Fmoc-Nle-OHFMOC-L-Norleucine

APOSOR480632-25GEA 74.6 GBP
APOSOR480632-25G
N-Fmoc-L-norleucine 98%
N-Fmoc-L-norleucine
Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037537
CAS Number: 77284-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR