4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radicalTempol4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl

APOSOR480623-250GEA 148 GBP
APOSOR480623-250G
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) 97%
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-Hydroxy-TEMPO, TEMPOL, free radical)
Formula: C₉H₁₈NO₂
MW: 172.25 g/mol
Melting Pt: 69…72 °C
Flash Pt: 146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006478
CAS Number: 2226-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR