L-Glutamic acid 5-benzyl ester 97%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: gamma-Benzyl L-glutamate5-Benzyl L-GlutamateH-Glu(OBzl)-OHγ-Benzyl-L-Glutamate

APOSOR480330-100GEA 59.9 GBP
APOSOR480330-100G
L-Glutamic acid 5-benzyl ester 97%
L-Glutamic acid 5-benzyl ester
Formula: C₁₂H₁₅NO₄
MW: 237.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002633
CAS Number: 1676-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR