2-Methylbenzothiazole 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR480326-100G OR480326-500G
APOSOR480326-100GEA 62 GBP
APOSOR480326-100G APOSOR480326-500G
2-Methylbenzothiazole 98%
2-Methylbenzothiazole
Formula: C₈H₇NS
MW: 149.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005794
CAS Number: 120-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR