4'-Amino-3',5'-dichloroacetophenone
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4′-Amino-3′,5′-dichloroacetophenone
APOSOR350012-100GEA
120
GBP
APOSOR350012-100G
4'-Amino-3',5'-dichloroacetophenone
4'-Amino-3',5'-dichloroacetophenone
Formula:
C₈H₇Cl₂NO MW: 204.0500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00238535 CAS Number: 37148-48-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...