2,4-Diamino-6-phenyl-1,3,5-triazine

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Phenyl-1,3,5-triazine-2,4-diyldiamine6-Phenyl-1,3,5-triazine-2,4-diamine2,4-Diamino-6-phenyl-s-triazineBenzoguanamine2,6-Diamino-4-phenyl-1,3,5-triazine

APOSOR346121-10GEA 0 GBP
APOSOR346121-10G APOSOR346121-5G APOSOR346121-1G
2,4-Diamino-6-phenyl-1,3,5-triazine
2,4-Diamino-6-phenyl-1,3,5-triazine
Formula: C₉H₉N₅
MW: 187.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023187
CAS Number: 91-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR