1-[3-(2-Pyrimidinyloxy)phenyl]-1-ethanone 95%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR33254-1G
OR33254-5G
APOSOR33254-1GEA
243
GBP
APOSOR33254-1G
APOSOR33254-5G
1-[3-(2-Pyrimidinyloxy)phenyl]-1-ethanone 95%
1-[3-(2-Pyrimidinyloxy)phenyl]-1-ethanone
Formula:
C₁₂H₁₀N₂O₂ MW: 214.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01568700 CAS Number: 339105-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...