5-Amino-1-(4-chlorophenyl)-3-methyl-1H-pyrazole 95%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(4-Chlorophenyl)-3-methyl-1H-pyrazol-5-amine

APOSOR32926-1GEA 316 GBP
APOSOR32926-1G
5-Amino-1-(4-chlorophenyl)-3-methyl-1H-pyrazole 95%
5-Amino-1-(4-chlorophenyl)-3-methyl-1H-pyrazole
Formula: C₁₀H₁₀ClN₃
MW: 207.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03478180
CAS Number: 40401-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR