Methanesulphonic acid 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

APOSOR322096-100GEA 19 GBP
APOSOR322096-100G APOSOR322096-500G
Methanesulphonic acid 99%
Methanesulphonic acid
Formula: CH₃SO₃H
MW: 96.11 g/mol
Boiling Pt: 167 °C/10mmHg
Melting Pt: 17…19 °C
Density: 1.481 g/cm³ (25 °C)
Flash Pt: 189 °C
Storage Temperature: Ambient
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 200-898-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR