Sodium-2-methyl-2-(pyrazin-2-yl)propanoate 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR318124-1G
APOSOR318124-1GEA 804 GBP
APOSOR318124-1G
Sodium-2-methyl-2-(pyrazin-2-yl)propanoate 95+%
Sodium-2-methyl-2-(pyrazin-2-yl)propanoate
Formula: C₈H₉N₂NaO₂
MW: 188.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12401611
CAS Number: 1208847-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR