Bromoferrocene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

APOSOR315061-5GEA 0 GBP
APOSOR315061-5G APOSOR315061-1G
Bromoferrocene
Bromoferrocene
Formula: C₁₀H₉BrFe
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD03093913
CAS Number: 1273-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR