2-Chloro-4,6-dimethoxy-1,3,5-triazine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Chloro-4,6-dimethoxy-s-triazine

OR307158-100G OR307158-500G
APOSOR307158-100GEA 52.5 GBP
APOSOR307158-100G APOSOR307158-500G
2-Chloro-4,6-dimethoxy-1,3,5-triazine 98%
2-Chloro-4,6-dimethoxy-1,3,5-triazine
Formula: C₅H₆ClN₃O₂
MW: 175.57 g/mol
Boiling Pt: 99…100 °C/1mmHg
Melting Pt: 71…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00075607
CAS Number: 3140-73-6
EINECS: 221-541-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR