2-(Tributylstannyl)thiophene
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
OR25730-100G
OR25730-10G
OR25730-50G
APOSOR25730-100GEA
147
GBP
APOSOR25730-100G
APOSOR25730-10G
APOSOR25730-50G
2-(Tributylstannyl)thiophene
2-(Tributylstannyl)thiophene
Formula:
C₁₆H₃₀SSn MW: 373.16 g/mol Boiling Pt: 155 °C/0.1mmHg Density: 1.175 g/cm³ (25 °C) Flash Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192513 CAS Number: 54663-78-4 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...