3-(Tributylstannyl)pyridine
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
3-(Tri-n-butylstannyl)pyridine
OR25463-1G
OR25463-25G
OR25463-5G
APOSOR25463-1GEA
94.9
GBP
APOSOR25463-1G
APOSOR25463-25G
APOSOR25463-5G
3-(Tributylstannyl)pyridine
3-(Tributylstannyl)pyridine
Formula:
C₁₇H₃₁NSn MW: 368.15 g/mol Boiling Pt: 140…146 °C/0.7mmHg Density: 1.153 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Freezer |
MDL Number:
MFCD00052054 CAS Number: 59020-10-9 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...