2-Acetyl-3-amino-5-tert-butylthiophene

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

APOSOR24302-100MGEA 21.1 GBP
APOSOR24302-100MG
2-Acetyl-3-amino-5-tert-butylthiophene
2-Acetyl-3-amino-5-tert-butylthiophene
Formula: C₁₀H₁₅NOS
Storage Temperature: Refrigerator
MDL Number: MFCD00068131
CAS Number: 175137-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR