Cupferron
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ammonium 2-oxo-1-phenylhydrazinolate
APOSOR2350-25GEA
45.3
GBP
APOSOR2350-25G
Cupferron
Cupferron
Formula:
C₆H₉N₃O₂ MW: 155.16 g/mol Melting Pt: 154 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00078422 CAS Number: 135-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...