4'-Hydroxybutyrophenone

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR22098-100G OR22098-25G OR22098-5G
APOSOR22098-100GEA 35.8 GBP
APOSOR22098-100G APOSOR22098-25G APOSOR22098-5G
4'-Hydroxybutyrophenone
4'-Hydroxybutyrophenone
Formula: C₁₀H₁₂O₂
Boiling Pt: 148…150 °C/11.5mmHg
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00020173
CAS Number: 1009-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR