(4-Aminophenyl)acetonitrile

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-(4-Aminophenyl)acetonitrile4-Aminobenzyl cyanide

APOSOR21111-1GEA 10.6 GBP
APOSOR21111-1G
(4-Aminophenyl)acetonitrile
(4-Aminophenyl)acetonitrile
Formula: C₈H₈N₂
MW: 132.17 g/mol
Boiling Pt: 312 °C
Melting Pt: 45…48 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007912
CAS Number: 3544-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR