N-Boc-2-chloroethylamine
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Chloroethylamine, N-BOC protected
OR18557-1G
OR18557-5G
APOSOR18557-1GEA
15.8
GBP
APOSOR18557-1G
APOSOR18557-5G
N-Boc-2-chloroethylamine
N-Boc-2-chloroethylamine
Formula:
C₇H₁₄ClNO₂ MW: 179.6446 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04004074 CAS Number: 71999-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...