Benzothiazol-2-ol 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Hydroxybenzothiazole
APOSOR16606-100GEA
77.8
GBP
APOSOR16606-100G
Benzothiazol-2-ol 98%
Benzothiazol-2-ol
Formula:
C₇H₅NOS MW: 151.18 g/mol Melting Pt: 137…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022868 CAS Number: 934-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...