2-Chloro-6-iodoaniline 98+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OR15148-1G
APOSOR15148-1GEA 26.3 GBP
APOSOR15148-1G
2-Chloro-6-iodoaniline 98+%
2-Chloro-6-iodoaniline
Formula: C₆H₅ClIN
MW: 253.4681 g/mol
Melting Pt: 64…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08443948
CAS Number: 84483-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR