4-Biphenylboronic acid pinacol ester 98%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
APOSOR15024-1GEA
10.6
GBP
APOSOR15024-1G
4-Biphenylboronic acid pinacol ester 98%
4-Biphenylboronic acid pinacol ester
Formula:
C₁₈H₂₁BO₂ MW: 280.17 g/mol Melting Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368291 CAS Number: 144432-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...