4-Bromo-L-phenylalanine 95+%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: H-Phe(4-Br)-OH

APOSOR14695-10GEA 66.2 GBP
APOSOR14695-10G
4-Bromo-L-phenylalanine 95+%
4-Bromo-L-phenylalanine
Formula: C₉H₁₀BrNO₂
MW: 244.1 g/mol
Melting Pt: 265 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063062
CAS Number: 24250-84-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR