4-Chloro-D-phenylalanine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: H-D-Phe(4-Cl)-OHH-p-Chloro-D-Phe-OH(R)-2-Amino-3-(4-chlorophenyl)propionic acid

APOSOR14692-1GEA 10.6 GBP
APOSOR14692-1G
4-Chloro-D-phenylalanine 98%
4-Chloro-D-phenylalanine
Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
Melting Pt: 260 °C
Storage Temperature: Freezer
MDL Number: MFCD00079675
CAS Number: 14091-08-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR