N,N'-Diphenylbenzidine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,4'-Bis(phenylamino)biphenyl, N4,N4'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine

OR11178-100G
APOSOR11178-100GEA 113 GBP
APOSOR11178-100G
N,N'-Diphenylbenzidine 98%
N,N'-Diphenylbenzidine
Formula: C₂₄H₂₀N₂
MW: 336.44 g/mol
Melting Pt: 247 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003016
CAS Number: 531-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR