Cyclooctanone 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OR10093-100G OR10093-500G
APOSOR10093-100GEA 70.4 GBP
APOSOR10093-100G APOSOR10093-500G
Cyclooctanone 98%
Cyclooctanone
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 195…197 °C
Melting Pt: 37…41 °C
Density: 0.958 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00001754
CAS Number: 502-49-8
EINECS: 207-940-2
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR