1-Cyclohexylpiperazine 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

APOSOR0872-250MGEA 10.6 GBP
APOSOR0872-250MG
1-Cyclohexylpiperazine 98%
1-Cyclohexylpiperazine
Formula: C₁₀H₂₀N₂
MW: 168.28 g/mol
Melting Pt: 32…34 °C
Storage Temperature: Ambient
MDL Number: MFCD00044809
CAS Number: 17766-28-8
EINECS: 241-750-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR