3-Methoxythiophenol
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
3-Methoxybenzenethiol, 1-Methoxy-3-mercaptobenzene, 3-Mercaptoanisol, 3-Methoxybenzene-1-thiol, 3-Methoxyphenyl mercaptan, m-Methoxythiophenol
OR0120-25G
APOSOR0120-25GEA
107
GBP
APOSOR0120-25G
3-Methoxythiophenol
3-Methoxythiophenol
Formula:
C₇H₈OS MW: 140.21 g/mol Boiling Pt: 225…226 °C Density: 1.136 g/cm³ (25 °C) Flash Pt: 96 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004841 CAS Number: 15570-12-4 EINECS: 239-617-7 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...