4-Nitro-3-picoline-N-oxide 99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Methyl-4-nitropyridine N-oxide4-Nitro-3-picoline N-oxide4-Nitropicoline N-oxide3-Methyl-4-nitropyridine 1-oxide3-Methyl-4-nitropyridine-1-oxide3-Methyl-4-nitropyridine-N-oxide

APOSOR0074-5GEA 10.6 GBP
APOSOR0074-5G
4-Nitro-3-picoline-N-oxide 99%
4-Nitro-3-picoline N-oxide
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00014626
CAS Number: 1074-98-2
EINECS: 214-050-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR