Potassium carbonate 99.99%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Potash

APOSIN2896-25GEA 73.6 GBP
APOSIN2896-25G
Potassium carbonate 99.99%
Potassium carbonate
Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891 °C
Density: 2.43 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR