Magnesium acetate tetrahydrate 99.999%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Magnesium di(acetate) tetrahydrateAcetic acid magnesium salt tetrahydrate

APOSIN2443-25GEA 174 GBP
APOSIN2443-25G
Magnesium acetate tetrahydrate 99.999%
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80 °C
Density: 1.454 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR