Methanol-[D₄] (>99,8% D)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Carbinol-D4Methyl alcohol-D4Methanol-[²H₄]Methyl-D3 alcohol-DWood alcohol-D4Methanol-d4Hydroxy methane-D4Tetradeuteromethanol

APOSDE70J-50MLEA 132 GBP
APOSDE70J-50ML
Methanol-[D₄] (>99,8% D)
Methanol-[D4]
Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.4 °C
Density: 0.89 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3+6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR