Tentoxin (from Alternaria tenuis)

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: cyclo-(N-Methyl-L-alanyl-L-leucyl-N-methyl-(Z)-dihydrophenylalanyl-glycyl)

BIT1002-1MG BIT1002-100UG
APOSBIT1002-1MGEA 558 GBP
APOSBIT1002-1MG APOSBIT1002-100UG
Tentoxin (from Alternaria tenuis)
Tentoxin
Formula: C₂₂H₃₀N₄O₄
MW: 414.5 g/mol
Storage Temperature: Refrigerator
CAS Number: 28540-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR